Assay Design



Gene Transcripts

Gene Symbol
Acot12
Entrez Gene ID
74156
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
1300004O04Rik, 4930449F15Rik, AV027244, Cach, mCACH-1
Chromosome Location
Chr.13: 91741511 - 91786148 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028790.3 NP_083066.1 12 - 13 1270 70
GenBank mRNA AB078618.1 - 12 - 13 1267 70
AK004905.1 - 12 - 13 1271 70
AK015431.1 - 2 - 3 248 70
AK033663.1 - 12 - 13 1270 70
AK034622.1 - 12 - 13 1274 70
BC025852.1 - 12 - 13 1223 70
BC132050.1 - 12 - 13 1301 70
BC137996.1 - 12 - 13 1301 70

View Full Product Details