Assay Design



Gene Transcripts

Gene Symbol
Higd1b
Entrez Gene ID
75689
Gene Name
HIG1 domain family, member 1B
Gene Aliases
2310056K19Rik
Chromosome Location
Chr.11: 102835756 - 102844133 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080846.1 NP_543122.1 1 - 2 472 97
GenBank mRNA AK009942.1 - 1 - 2 313 97
BC005568.1 - 1 - 2 472 97

View Full Product Details