Assay Design



Gene Transcripts

Gene Symbol
Rogdi
Entrez Gene ID
66049
Gene Name
rogdi homolog (Drosophila)
Gene Aliases
0610011C19Rik, AU020118, C76152, Lzf
Chromosome Location
Chr.16: 5008729 - 5013805 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133185.2 NP_573448.2 8 - 9 690 130
GenBank mRNA AK002523.1 - 8 - 9 131
AK170144.1 - 8 - 9 655 130
AY036117.1 - 8 - 9 658 130
BC006914.1 - 8 - 9 733 130
CD363552.1 - 3 - 4 256 130

View Full Product Details