Assay Design



Gene Transcripts

Gene Symbol
Cux2
Entrez Gene ID
13048
Gene Name
cut-like homeobox 2
Gene Aliases
1700051K22Rik, Cutl2, Cux-2, ENSMUSG00000072641
Chromosome Location
Chr.5: 121857962 - 122050562 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007804.2 NP_031830.2 5 - 6 566 68
GenBank mRNA BC065113.1 - 5 - 6 566 68
U45665.1 - 5 - 6 387 68

View Full Product Details