Assay Design



Gene Transcripts

Gene Symbol
Gtf2h1
Entrez Gene ID
14884
Gene Name
general transcription factor II H, polypeptide 1
Gene Aliases
62kDa, AW743425, AW822074, BTF2 p62, C77871, p62
Chromosome Location
Chr.7: 46795812 - 46823800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291075.1 NP_001278004.1 4 - 5 521 113
NM_008186.4 NP_032212.3 3 - 4 554 113
GenBank mRNA BC052837.1 - 4 - 5 533 113

View Full Product Details