Assay Design



Gene Transcripts

Gene Symbol
Map7
Entrez Gene ID
17761
Gene Name
microtubule-associated protein 7
Gene Aliases
E-MAP-115, MAP-7, Mtap7, R75000, mshi, mste, ste
Chromosome Location
Chr.10: 20148920 - 20281590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008635.2 NP_032661.2 7 - 8 842 79
GenBank mRNA AB098611.1 - 7 - 8 748 79
AK014575.1 - 7 - 8 759 79
AK133267.1 - 7 - 8 1291 79
BC042771.1 - 7 - 8 767 79
BC044733.1 - 7 - 8 767 79
BC046555.1 - 7 - 8 767 79
Y15197.1 - 7 - 8 748 79

View Full Product Details