Assay Design



Gene Transcripts

Gene Symbol
Ncoa3
Entrez Gene ID
17979
Gene Name
nuclear receptor coactivator 3
Gene Aliases
2010305B15Rik, AW321064, Actr, Aib1, KAT13B, Rac3, Src3, Tram-1, Tram1, bHLHe42, p/Cip, pCip
Chromosome Location
Chr.2: 165992637 - 166073242 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008679.3 NP_032705.2 14 - 15 2895 89
GenBank mRNA AF000581.1 - 13 - 14 2787 89
AK142306.1 - 14 - 15 2800 89
AK156345.1 - 14 - 15 2872 89
BC086482.1 - 14 - 15 2865 89
BC093478.1 - 11 - 12 2526 89
BC141177.1 - 13 - 14 2726 89

View Full Product Details