Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113209.2 | NP_001106680.1 | 1 - 2 | 1052 | 92 |
NM_001113210.2 | NP_001106681.1 | 1 - 2 | 1052 | 92 | |
NM_001286127.1 | NP_001273056.1 | 1 - 2 | 1052 | 92 | |
NM_001286131.1 | NP_001273060.1 | 1 - 2 | 1052 | 92 | |
NM_008687.6 | NP_032713.3 | 1 - 2 | 1052 | 92 | |
GenBank mRNA | AK034261.1 | - | 1 - 2 | 584 | 92 |
AK034793.1 | - | 1 - 2 | 551 | 92 | |
AK047576.1 | - | 1 - 2 | 1053 | 92 | |
AK083167.1 | - | 1 - 2 | 397 | 92 | |
AK084941.1 | - | 1 - 2 | 92 | ||
AK087480.1 | - | 1 - 2 | 584 | 92 | |
AK146480.1 | - | 1 - 2 | 569 | 92 | |
AK148573.1 | - | 1 - 2 | 1040 | 92 | |
AY035852.1 | - | 1 - 2 | 57 | 92 | |
AY035853.1 | - | 1 - 2 | 57 | 92 | |
BC014290.1 | - | 1 - 2 | 89 | 92 | |
BC023366.1 | - | 1 - 2 | 189 | 92 | |
BC062908.1 | - | 1 - 2 | 374 | 92 | |
U57634.1 | - | 1 - 2 | 258 | 92 | |
Y07685.1 | - | 1 - 2 | 984 | 92 | |
Y07686.1 | - | 1 - 2 | 564 | 92 | |
Y07687.1 | - | 1 - 2 | 1045 | 92 |