Assay Design



Gene Transcripts

Gene Symbol
Npas2
Entrez Gene ID
18143
Gene Name
neuronal PAS domain protein 2
Gene Aliases
MOP4, bHLHe9
Chromosome Location
Chr.1: 39193956 - 39363240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008719.2 NP_032745.2 5 - 6 858 86
GenBank mRNA BC109166.2 - 5 - 6 858 86
U77969.1 - 5 - 6 932 86

View Full Product Details