Assay Design



Gene Transcripts

Gene Symbol
Ptpn22
Entrez Gene ID
19260
Gene Name
protein tyrosine phosphatase, non-receptor type 22 (lymphoid)
Gene Aliases
70zpep, PEP, Ptpn8
Chromosome Location
Chr.3: 103856804 - 103912252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008979.2 NP_033005.1 8 - 9 814 114
NR_104070.1 10 - 11 839 114
GenBank mRNA AK153812.1 - 10 - 11 840 114
BC055377.1 - 8 - 9 736 114
M90388.1 - 8 - 9 799 114

View Full Product Details