Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042541.1 | NP_001036006.1 | 2 - 3 | 1802 | 71 |
NM_009648.2 | NP_033778.2 | 2 - 3 | 1765 | 71 | |
GenBank mRNA | AK076216.1 | - | 2 - 3 | 1804 | 71 |
AK132749.1 | - | 2 - 3 | 1776 | 71 | |
AK196354.1 | - | 1 - 2 | 139 | 71 | |
BC065135.1 | - | 2 - 3 | 1714 | 71 | |
U95146.1 | - | 2 - 3 | 1778 | 71 |