Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146038.2 | NP_001139510.1 | 2 - 3 | 292 | 115 |
NM_001271627.1 | NP_001258556.1 | 1 - 2 | 456 | 115 | |
NM_009820.5 | NP_033950.2 | 2 - 3 | 259 | 115 | |
GenBank mRNA | AF005936.1 | - | 1 - 2 | 410 | 115 |
AF010284.1 | - | 2 - 3 | 259 | 115 |