Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009951.4 | NP_034081.1 | 14 - 15 | 1954 | 88 |
GenBank mRNA | AF061569.1 | - | 14 - 15 | 1773 | 88 |
AK013940.1 | - | 14 - 15 | 1921 | 88 | |
AK044850.1 | - | 14 - 15 | 1859 | 88 | |
BC051679.1 | - | 14 - 15 | 1937 | 88 |