Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010848.3 | NP_034978.3 | 9 - 10 | 1470 | 104 |
GenBank mRNA | AK088598.1 | - | 9 - 10 | 1392 | 104 |
AK132250.1 | - | 9 - 10 | 1335 | 104 | |
AK137794.1 | - | 3 - 4 | 415 | 104 | |
AK137965.1 | - | 9 - 10 | 1336 | 104 | |
BC011513.1 | - | 9 - 10 | 1378 | 104 | |
M12848.1 | - | 9 - 10 | 1468 | 104 |