Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202445.1 | NP_001189374.1 | 8 - 9 | 2197 | 75 |
NM_001202446.1 | NP_001189375.1 | 8 - 9 | 2122 | 75 | |
NM_011162.5 | NP_035292.2 | 8 - 9 | 1905 | 75 | |
GenBank mRNA | AB221603.1 | - | 8 - 9 | 1768 | 75 |
AF054611.1 | - | 8 - 9 | 1905 | 75 | |
AF109768.1 | - | 8 - 9 | 1802 | 78 | |
AF109769.1 | - | 8 - 9 | 1865 | 78 | |
AF109770.1 | - | 8 - 9 | 1920 | 78 | |
AF109771.1 | - | 8 - 9 | 1766 | 78 | |
AF332075.1 | - | 8 - 9 | 1768 | 75 | |
AF332076.1 | - | 8 - 9 | 1768 | 75 | |
AK147033.1 | - | 8 - 9 | 2124 | 75 | |
BC072578.1 | - | 8 - 9 | 2089 | 75 |