Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011465.4 | NP_035595.2 | 51 - 52 | 7285 | 119 |
GenBank mRNA | AF093576.1 | - | 51 - 52 | 7193 | 119 |
AK086017.1 | - | 8 - 9 | 925 | 119 | |
AK143823.1 | - | 15 - 16 | 1953 | 119 | |
AK161449.1 | - | 21 - 22 | 4211 | 119 | |
BC150747.1 | - | 51 - 52 | 7150 | 119 | |
U87455.1 | - | 51 - 52 | 7265 | 119 |