Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080118.1 | NP_001073587.1 | 14 - 15 | 1508 | 105 |
NM_013634.2 | NP_038662.2 | 14 - 15 | 1373 | 105 | |
NM_134027.2 | NP_598788.2 | 14 - 15 | 1508 | 105 | |
GenBank mRNA | AF000294.1 | - | 14 - 15 | 1373 | 105 |
AF332073.1 | - | 13 - 14 | 1253 | 105 | |
AF332074.1 | - | 13 - 14 | 1253 | 105 | |
AK049203.1 | - | 14 - 15 | 1499 | 105 | |
AK147199.1 | - | 14 - 15 | 1485 | 105 | |
BC021440.1 | - | 14 - 15 | 1479 | 105 | |
BC079636.1 | - | 14 - 15 | 1478 | 105 |