Assay Design



Gene Transcripts

Gene Symbol
Phc2
Entrez Gene ID
54383
Gene Name
polyhomeotic-like 2 (Drosophila)
Gene Aliases
A3galt2, AA415044, D130050K19Rik, D4Ertd810e, Edr2, Mph2, p36
Chromosome Location
Chr.4: 128654673 - 128752881 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195130.1 NP_001182059.1 3 - 4 514 107
NM_018774.4 NP_061244.1 3 - 4 507 107
GenBank mRNA BC057571.1 - 3 - 4 450 107
U81491.1 - 2 - 3 404 107

View Full Product Details