Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195083.1 | NP_001182012.1 | 6 - 7 | 1168 | 131 |
NM_001195130.1 | NP_001182059.1 | 14 - 15 | 2568 | 131 | |
NM_018774.4 | NP_061244.1 | 14 - 15 | 2561 | 131 | |
GenBank mRNA | AB062362.1 | - | 6 - 7 | 1154 | 131 |
AF060076.1 | - | 7 - 8 | 1147 | 131 | |
BC051657.1 | - | 6 - 7 | 1036 | 131 | |
BC057571.1 | - | 14 - 15 | 2504 | 131 | |
BC071246.1 | - | 6 - 7 | 1149 | 131 | |
U81491.1 | - | 13 - 14 | 2458 | 131 |