Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019764.2 | NP_062738.2 | 5 - 6 | 1425 | 60 |
GenBank mRNA | AF175967.1 | - | 5 - 6 | 708 | 60 |
AF175968.1 | - | 3 - 4 | 1565 | 60 | |
AK040912.1 | - | 5 - 6 | 1506 | 60 | |
AK077535.1 | - | 5 - 6 | 1421 | 60 | |
AK141348.1 | - | 5 - 6 | 1425 | 60 | |
AK164279.1 | - | 5 - 6 | 1494 | 60 | |
AK164819.1 | - | 5 - 6 | 1425 | 60 | |
AK169419.1 | - | 5 - 6 | 1425 | 60 | |
AX763686.1 | - | 4 - 5 | 1264 | 60 | |
BC027824.1 | - | 5 - 6 | 1395 | 60 | |
BC035201.1 | - | 5 - 6 | 1407 | 60 | |
BC108411.1 | - | 5 - 6 | 1459 | 60 |