Assay Design
Gene Transcripts
- Gene Symbol
- Rsrp1
- Entrez Gene ID
-
27981
- Gene Name
- arginine/serine rich protein 1
- Gene Aliases
-
2700043I21Rik, D4Ucla2, D4Wsu53e
- Chromosome Location
-
Chr.4: 134923625 - 134927370 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB041555.1 |
- |
3 - 4 |
|
139 |
AK169598.1 |
- |
3 - 4 |
892 |
141 |
View Full Product Details