Assay Design



Gene Transcripts

Gene Symbol
Fuca1
Entrez Gene ID
71665
Gene Name
fucosidase, alpha-L- 1, tissue
Gene Aliases
0610006A03Rik, 9530055J05Rik, Afuc, Fuca
Chromosome Location
Chr.4: 135920726 - 135940300 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024243.4 NP_077205.3 3 - 4 697 84
GenBank mRNA AK002230.1 - 3 - 4 665 84
AK053323.1 - 3 - 4 642 84
AK089958.1 - 3 - 4 667 84
AK150526.1 - 3 - 4 632 84
AK150890.1 - 3 - 4 687 84
AK151258.1 - 3 - 4 665 84
AK151361.1 - 3 - 4 686 84
AK151879.1 - 3 - 4 686 84
AK151908.1 - 3 - 4 640 84
AK152070.1 - 3 - 4 640 84
AK165604.1 - 3 - 4 667 84
AK170503.1 - 3 - 4 677 84
AK204459.1 - 2 - 3 191 84
BC003235.1 - 3 - 4 670 84

View Full Product Details