Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025411.4 | NP_079687.3 | 5 - 6 | 644 | 88 |
GenBank mRNA | AF400670.1 | - | 5 - 6 | 607 | 88 |
AK004208.1 | - | 5 - 6 | 645 | 88 | |
AK013877.1 | - | 3 - 4 | 307 | 88 | |
AK050261.1 | - | 5 - 6 | 617 | 88 | |
BC052695.1 | - | 5 - 6 | 544 | 88 |