Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026844.3 | NP_081120.2 | 1 - 2 | 86 | 66 |
GenBank mRNA | AK002429.1 | - | 1 - 2 | 86 | 66 |
AK004189.1 | - | 1 - 2 | 103 | 66 | |
AK009995.1 | - | 1 - 2 | 73 | 66 | |
AK010073.1 | - | 1 - 2 | 73 | 66 | |
BC027549.1 | - | 1 - 2 | 56 | 66 |