Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163661.1 | NP_001157133.1 | 10 - 11 | 1927 | 126 |
NM_027880.2 | NP_082156.2 | 10 - 11 | 1838 | 126 | |
GenBank mRNA | AK004582.1 | - | 10 - 11 | 1708 | 126 |
AK028658.1 | - | 10 - 11 | 1692 | 126 | |
AK045321.1 | - | 3 - 4 | 1374 | 126 | |
AK050773.1 | - | 10 - 11 | 1657 | 126 | |
AK084830.1 | - | 10 - 11 | 1708 | 126 | |
AK129189.1 | - | 9 - 10 | 2000 | 126 | |
AK143827.1 | - | 10 - 11 | 1928 | 126 | |
AK146963.1 | - | 10 - 11 | 1714 | 126 | |
AK154727.1 | - | 10 - 11 | 1740 | 126 | |
AK158181.1 | - | 11 - 12 | 126 | ||
BC011077.1 | - | 10 - 11 | 1688 | 126 | |
BC080713.1 | - | 9 - 10 | 1947 | 126 |