Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028238.7 | NP_082514.4 | 2 - 3 | 616 | 68 |
GenBank mRNA | AB232634.1 | - | 2 - 3 | 487 | 68 |
AK044783.1 | - | 2 - 3 | 579 | 68 | |
AK054253.1 | - | 2 - 3 | 617 | 68 | |
AK165682.1 | - | 2 - 3 | 619 | 68 | |
AK196794.1 | - | 1 - 2 | 247 | 68 | |
AK197116.1 | - | 1 - 2 | 247 | 68 | |
AY062237.1 | - | 2 - 3 | 576 | 68 | |
BC089578.1 | - | 2 - 3 | 576 | 68 |