Assay Design



Gene Transcripts

Gene Symbol
Plekha2
Entrez Gene ID
83436
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2
Gene Aliases
6430512N22Rik, AI851530, AU041791, TAPP2, W91773
Chromosome Location
Chr.8: 25039144 - 25104917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031257.3 NP_112547.1 4 - 5 425 94
GenBank mRNA AF286161.1 - 4 - 5 379 94
AF418551.1 - 3 - 4 291 94
AK032325.1 - 4 - 5 435 94
AK042324.1 - 4 - 5 399 94
AK078212.1 - 4 - 5 404 94
AK088395.1 - 4 - 5 367 94
AK153117.1 - 4 - 5 397 94
BC010215.1 - 4 - 5 346 94

View Full Product Details