Assay Design



Gene Transcripts

Gene Symbol
Plekha2
Entrez Gene ID
83436
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2
Gene Aliases
6430512N22Rik, AI851530, AU041791, TAPP2, W91773
Chromosome Location
Chr.8: 25039144 - 25104917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031257.3 NP_112547.1 6 - 7 662 61
GenBank mRNA AF286161.1 - 6 - 7 616 61
AF418551.1 - 5 - 6 528 61
AK042324.1 - 6 - 7 636 61
AK078212.1 - 6 - 7 641 61
AK088395.1 - 6 - 7 604 61
AK153117.1 - 6 - 7 634 61
BC010215.1 - 6 - 7 583 61

View Full Product Details