Assay Design
Gene Transcripts
- Gene Symbol
- Sncb
- Entrez Gene ID
-
104069
- Gene Name
- synuclein, beta
- Gene Aliases
-
AI838531, betaSYN
- Chromosome Location
-
Chr.13: 54758860 - 54766440 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_033610.2 |
NP_291088.1 |
5 - 6 |
516 |
81 |
GenBank mRNA |
BC019409.1 |
- |
5 - 6 |
516 |
81 |
View Full Product Details