Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053262.3 | NP_444492.1 | 5 - 6 | 750 | 88 |
GenBank mRNA | AF304306.1 | - | 5 - 6 | 746 | 88 |
AK154803.1 | - | 5 - 6 | 765 | 88 | |
AK155174.1 | - | 5 - 6 | 765 | 88 | |
AK155202.1 | - | 5 - 6 | 770 | 88 | |
AK155327.1 | - | 5 - 6 | 770 | 88 | |
AK158327.1 | - | 5 - 6 | 765 | 88 | |
AK170939.1 | - | 5 - 6 | 765 | 88 | |
AY053570.1 | - | 5 - 6 | 764 | 88 | |
BC038340.1 | - | 5 - 6 | 750 | 88 |