Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039512.1 | NP_001034601.1 | 11 - 12 | 1749 | 81 |
NM_054102.2 | NP_473443.2 | 12 - 13 | 1875 | 81 | |
GenBank mRNA | AB055737.1 | - | 12 - 13 | 1871 | 81 |
AK129229.1 | - | 12 - 13 | 1813 | 81 | |
AK144619.1 | - | 12 - 13 | 1855 | 81 | |
AK144835.1 | - | 11 - 12 | 1730 | 81 | |
AK145071.1 | - | 12 - 13 | 1866 | 81 | |
AK145845.1 | - | 12 - 13 | 1825 | 81 | |
AK146001.1 | - | 12 - 13 | 1856 | 81 | |
AK146494.1 | - | 12 - 13 | 1828 | 81 | |
AK146645.1 | - | 12 - 13 | 1826 | 81 | |
AK159254.1 | - | 12 - 13 | 1857 | 81 | |
BC004092.1 | - | 11 - 12 | 1575 | 81 | |
DQ914522.1 | - | 12 - 13 | 1707 | 81 | |
DQ914523.1 | - | 12 - 13 | 2758 | 81 |