Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_179203.3 | NP_849534.2 | 12 - 13 | 1355 | 82 |
GenBank mRNA | AF343079.1 | - | 12 - 13 | 1401 | 82 |
AK075921.1 | - | 12 - 13 | 1351 | 82 | |
AK148974.1 | - | 12 - 13 | 1354 | 82 | |
AK149702.1 | - | 12 - 13 | 1354 | 82 | |
AK150037.1 | - | 12 - 13 | 1354 | 82 | |
AK165724.1 | - | 12 - 13 | 1353 | 82 | |
AK166099.1 | - | 12 - 13 | 1351 | 82 | |
AK172038.1 | - | 12 - 13 | 1351 | 82 | |
AK172350.1 | - | 12 - 13 | 1356 | 82 | |
AK218475.1 | - | 2 - 3 | 108 | 82 | |
BC023301.1 | - | 12 - 13 | 1292 | 82 | |
BC058373.1 | - | 12 - 13 | 1339 | 82 | |
BC060036.1 | - | 12 - 13 | 1287 | 82 |