Assay Design



Gene Transcripts

Gene Symbol
Qprt
Entrez Gene ID
67375
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
2410027J01Rik, AI647766, QPRTase
Chromosome Location
Chr.7: 127107770 - 127122029 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133686.1 NP_598447.1 1 - 2 53 70
GenBank mRNA AK010602.1 - 1 - 2 69 70
AK019119.1 - 1 - 2 69 70
BC011191.1 - 1 - 2 53 70

View Full Product Details