Assay Design



Gene Transcripts

Gene Symbol
Ppp1r32
Entrez Gene ID
67752
Gene Name
protein phosphatase 1, regulatory subunit 32
Gene Aliases
4930579J09Rik, AU015816, IIIG9, IIIG9L, IIIG9S
Chromosome Location
Chr.19: 10474235 - 10491452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133689.1 NP_598450.1 1 - 2 73 83
GenBank mRNA AK005821.1 - 1 - 2 105 83
AY032666.1 - 1 - 2 73 83
BC118523.1 - 1 - 2 49 83

View Full Product Details