Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133689.1 | NP_598450.1 | 8 - 9 | 845 | 86 |
GenBank mRNA | AK005821.1 | - | 8 - 9 | 878 | 86 |
AK016325.1 | - | 8 - 9 | 873 | 86 | |
AY032666.1 | - | 8 - 9 | 845 | 86 | |
BC118523.1 | - | 8 - 9 | 821 | 86 |