Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164600.1 | NP_001158072.1 | 4 - 5 | 359 | 108 |
NM_133691.5 | NP_598452.2 | 4 - 5 | 389 | 108 | |
GenBank mRNA | AK075960.1 | - | 4 - 5 | 357 | 108 |
AK151526.1 | - | 4 - 5 | 353 | 108 | |
AK168789.1 | - | 4 - 5 | 356 | 108 | |
BC010601.1 | - | 4 - 5 | 359 | 108 |