Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164600.1 | NP_001158072.1 | 9 - 10 | 1150 | 87 |
NM_028364.2 | NP_082640.2 | 10 - 11 | 1231 | 87 | |
NM_133691.5 | NP_598452.2 | 9 - 10 | 1180 | 87 | |
GenBank mRNA | AK012434.1 | - | 9 - 10 | 1118 | 87 |
AK075960.1 | - | 9 - 10 | 1148 | 87 | |
AK146534.1 | - | 10 - 11 | 1202 | 87 | |
AK149632.1 | - | 10 - 11 | 1197 | 87 | |
AK151526.1 | - | 9 - 10 | 1144 | 87 | |
AK168789.1 | - | 9 - 10 | 1147 | 87 | |
BC010601.1 | - | 9 - 10 | 1150 | 87 |