Assay Design



Gene Transcripts

Gene Symbol
Lancl2
Entrez Gene ID
71835
Gene Name
LanC (bacterial lantibiotic synthetase component C)-like 2
Gene Aliases
1700003F10Rik
Chromosome Location
Chr.6: 57702455 - 57739449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133737.2 NP_598498.1 1 - 2 804 84
GenBank mRNA AB035967.1 - 1 - 2 623 84
AK005632.1 - 1 - 2 173 84
BC016072.1 - 1 - 2 658 84

View Full Product Details