Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042652.1 | NP_001036117.1 | 2 - 3 | 674 | 93 |
NM_133851.3 | NP_598612.1 | 2 - 3 | 674 | 93 | |
GenBank mRNA | AF305710.1 | - | 2 - 3 | 255 | 93 |
AK042697.1 | - | - | 255 | 93 | |
AK076028.1 | - | 2 - 3 | 244 | 93 | |
AK088389.1 | - | 2 - 3 | 244 | 93 | |
BC004787.1 | - | 2 - 3 | 224 | 93 | |
BC100392.1 | - | 2 - 3 | 674 | 93 |