Assay Design



Gene Transcripts

Gene Symbol
Nfxl1
Entrez Gene ID
100978
Gene Name
nuclear transcription factor, X-box binding-like 1
Gene Aliases
1700012H24Rik, AW538212, D430033A06Rik, DNABF, GCF, Gm1058, TCF9
Chromosome Location
Chr.5: 72513301 - 72559707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133921.2 NP_598682.2 19 - 20 2398 73
GenBank mRNA AK132320.1 - 18 - 19 2308 73
BC022652.1 - 3 - 4 398 73

View Full Product Details