Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134025.3 | NP_598786.1 | 3 - 4 | 933 | 80 |
GenBank mRNA | AK035552.1 | - | 3 - 4 | 934 | 80 |
AK041175.1 | - | 3 - 4 | 1029 | 80 | |
AK153798.1 | - | 2 - 3 | 1140 | 80 | |
AK166413.1 | - | 3 - 4 | 977 | 80 | |
BC021800.1 | - | 2 - 3 | 757 | 80 |