Assay Design



Gene Transcripts

Gene Symbol
Cyp4f15
Entrez Gene ID
106648
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 15
Gene Aliases
AI787289, Cyp4f-15
Chromosome Location
Chr.17: 32685601 - 32703352 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134127.1 NP_598888.1 11 - 12 1486 88
GenBank mRNA AF233645.1 - 11 - 12 1486 88
BC021377.1 - 11 - 12 1456 88

View Full Product Details