Product Details
- Assay Reference Genome
Location
Chr.19:44761752 on build GRCm38
- Cytoband
- 19C3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 18504
- Gene Symbol
- Pax2
- Gene Name
- paired box 2
- Gene Aliases
- Opdc, Pax-2
- Location
- Chr.19:44745695-44837871 on build GRCm38
- Assay Gene Location
- Within Exon 3
View Full Product Details