Assay Design



Gene Transcripts

Gene Symbol
Ogt
Entrez Gene ID
108155
Gene Name
O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase)
Gene Aliases
1110038P24Rik, 4831420N21Rik, AI115525, Ogtl
Chromosome Location
Chr.X: 101640012 - 101684351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290535.1 NP_001277464.1 7 - 8 961 90
NM_139144.4 NP_631883.2 7 - 8 1158 90
GenBank mRNA AF363030.1 - 7 - 8 923 90
BC038435.1 - 3 - 4 1217 90
BC048239.1 - 3 - 4 914 90
BC057319.1 - 7 - 8 1081 90

View Full Product Details