Assay Design



Gene Transcripts

Gene Symbol
Spag4
Entrez Gene ID
245865
Gene Name
sperm associated antigen 4
Gene Aliases
1700041K21Rik, MNCb-0953, Sun4, mKIAA4118
Chromosome Location
Chr.2: 156065015 - 156069501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139151.4 NP_631890.3 10 - 11 1231 60
GenBank mRNA AB041554.1 - 8 - 9 1810 60
AK220494.1 - - 2504 60

View Full Product Details