Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144522.5 | NP_653105.3 | 1 - 2 | 1011 | 78 |
GenBank mRNA | AF285112.1 | - | 1 - 2 | 369 | 78 |
AK047320.1 | - | 1 - 2 | 1012 | 78 | |
AK047327.1 | - | 1 - 2 | 1012 | 78 | |
BC072576.1 | - | 1 - 2 | 554 | 78 |