Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020033.1 | NP_064417.1 | 8 - 9 | 850 | 103 |
GenBank mRNA | AB076256.1 | - | 8 - 9 | 862 | 103 |
AJ011118.1 | - | 8 - 9 | 103 | ||
AJ245514.1 | - | 8 - 9 | 850 | 103 | |
AK132585.1 | - | 8 - 9 | 860 | 103 | |
BC138760.1 | - | 8 - 9 | 888 | 103 | |
BC138761.1 | - | 8 - 9 | 888 | 103 |