Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039189.2 | NP_001034278.1 | 2 - 3 | 523 | 87 |
NM_024274.3 | NP_077236.1 | 3 - 4 | 1156 | 87 | |
GenBank mRNA | AK020164.1 | - | 2 - 3 | 309 | 87 |
AK076259.1 | - | 3 - 4 | 940 | 87 | |
AK170986.1 | - | 3 - 4 | 939 | 87 | |
BC002147.1 | - | 3 - 4 | 926 | 87 |