Assay Design



Gene Transcripts

Gene Symbol
Susd3
Entrez Gene ID
66329
Gene Name
sushi domain containing 3
Gene Aliases
1700017I11Rik, 2810440J20Rik
Chromosome Location
Chr.13: 49230827 - 49248706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025491.3 NP_079767.2 1 - 2 118 94
GenBank mRNA AK003860.1 - 1 - 2 118 94
AK006068.1 - 1 - 2 109 94
AK013276.1 - 1 - 2 110 94

View Full Product Details