Assay Design



Gene Transcripts

Gene Symbol
Susd3
Entrez Gene ID
66329
Gene Name
sushi domain containing 3
Gene Aliases
1700017I11Rik, 2810440J20Rik
Chromosome Location
Chr.13: 49230827 - 49248706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025491.3 NP_079767.2 4 - 5 642 60
NM_028340.1 NP_082616.1 4 - 5 811 60
GenBank mRNA AK003860.1 - 4 - 5 642 60
AK006068.1 - 4 - 5 632 60
BC045525.1 - 4 - 5 811 60

View Full Product Details